Ethyl-5-amino-1-(2-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-2314-1G
COBBHC-2314-1GEA 149 EUR
COBBHC-2314-1G
Ethyl-5-amino-1-(2-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Ethyl-5-amino-1-(2-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Formel: C₁₃H₁₁ClN₄O₂
MDL: MFCD09835739
CAS-Nummer: 1150164-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR