5,8-Difluorchinolin

Lieferant: COMBI-BLOCKS

HC-2330-1G
COBBHC-2330-1GEA 404 EUR
COBBHC-2330-1G
5,8-Difluorchinolin
5,8-Difluoroquinoline
Formel: C₉H₅F₂N
MDL: MFCD01939053
CAS-Nummer: 16650-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR