2-Brom-4-iodtoluol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-4-iodo-1-methylbenzene, 1-Bromo-5-iodo-2-metnylbenzene

HC-2382-5G
COBBHC-2382-5GEA 115 EUR
COBBHC-2382-5G
2-Brom-4-iodtoluol
2-Brom-4-iodtoluol
Formel: C₇H₆BrI
Molecular Weight: 296,9300 g/mol
MDL: MFCD07779014
CAS-Nummer: 26670-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR