Ethyl-5-amino-4-cyan-1-(4-fluorphenyl)-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 5-amino-4-cyano-1-(4-fluorophenyl)-1H-pyrazole-3-carboxylate

HC-2396-1G
COBBHC-2396-1GEA 149 EUR
COBBHC-2396-1G
Ethyl-5-amino-4-cyan-1-(4-fluorphenyl)-1H-pyrazol-3-carboxylat
Ethyl-5-amino-4-cyan-1-(4-fluorphenyl)-1H-pyrazol-3-carboxylat
Formel: C₁₃H₁₁FN₄O₂
Molecular Weight: 274,2505 g/mol
MDL: MFCD09835734
CAS-Nummer: 1001665-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR