Ethyl-5-amino-4-cyan-1-(3-fluorphenyl)-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-2397-1G
COBBHC-2397-1GEA 149 EUR
COBBHC-2397-1G
Ethyl-5-amino-4-cyan-1-(3-fluorphenyl)-1H-pyrazol-3-carboxylat
Ethyl-5-amino-4-cyan-1-(3-fluorphenyl)-1H-pyrazol-3-carboxylat
Formel: C₁₃H₁₁FN₄O₂
MDL: MFCD09835735
CAS-Nummer: 1150164-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR