Ethyl-5-amino-4-cyan-1-(2-fluorphenyl)-1H-pyrazol-3-carboxylat
Lieferant: COMBI-BLOCKS
HC-2398-1G
COBBHC-2398-1GEA
149
EUR
COBBHC-2398-1G
Ethyl-5-amino-4-cyan-1-(2-fluorphenyl)-1H-pyrazol-3-carboxylat
Ethyl-5-amino-4-cyan-1-(2-fluorphenyl)-1H-pyrazol-3-carboxylat
Formel:
C₁₃H₁₁FN₄O₂ |
MDL:
MFCD09835736 CAS-Nummer: 1150164-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...