Ethyl-5-amino-4-cyan-1-(4-methylphenyl)-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-2399-1G
COBBHC-2399-1GEA 149 EUR
COBBHC-2399-1G
Ethyl-5-amino-4-cyan-1-(4-methylphenyl)-1H-pyrazol-3-carboxylat
Ethyl-5-amino-4-cyan-1-(4-methylphenyl)-1H-pyrazol-3-carboxylat
Formel: C₁₄H₁₄N₄O₂
MDL: MFCD09835731
CAS-Nummer: 152992-58-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR