4-Chlor-6-methoxy-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2402-1G
COBBHC-2402-1GEA 0 EUR
COBBHC-2402-1G
4-Chlor-6-methoxy-2-(trifluormethyl)chinolin
4-Chloro-6-methoxy-2-(trifluoromethyl)quinoline
Formel: C₁₁H₇ClF₃NO
MDL: MFCD11855899
CAS-Nummer: 1701-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR