5-Nitro-6-(trifluormethoxy)chinolin

Lieferant: COMBI-BLOCKS

HC-2416-1G
COBBHC-2416-1GEA 98.3 EUR
COBBHC-2416-1G
5-Nitro-6-(trifluormethoxy)chinolin
5-Nitro-6-(trifluoromethoxy)quinoline
Formel: C₁₀H₅F₃N₂O₃
MDL: MFCD11855900
CAS-Nummer: 1133115-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR