1-(5-Nitrochinolin-6-yl)piperidin-4-ol

Lieferant: COMBI-BLOCKS

HC-2429-1G
COBBHC-2429-1GEA 0 EUR
COBBHC-2429-1G
1-(5-Nitrochinolin-6-yl)piperidin-4-ol
1-(5-Nitrochinolin-6-yl)piperidin-4-ol
Formel: C₁₄H₁₅N₃O₃
MDL: MFCD11855904
CAS-Nummer: 1133115-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR