1,3-Dichlor-5-iodbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dichloroiodobenzene

HC-2468-5G HC-2468-25G HC-2468-100G
COBBHC-2468-5GEA 25.1 EUR
COBBHC-2468-5G COBBHC-2468-25G COBBHC-2468-100G
1,3-Dichlor-5-iodbenzol
1,3-Dichlor-5-iodbenzol
Formel: C₆H₃Cl₂I
Molecular Weight: 272.9 g/mol
Siedepunkt: 127 °C (20 mmHg)
Schmelzpunkt: 57
MDL: MFCD00001047
CAS-Nummer: 3032-81-3
EINECS: 221-216-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR