Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Lieferant: COMBI-BLOCKS
HC-2500-5G
HC-2500-1G
HC-2500-250M
COBBHC-2500-5GEA
0
EUR
COBBHC-2500-5G
COBBHC-2500-1G
COBBHC-2500-250M
Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Formel:
C₉H₇BrN₂O₂ |
MDL:
MFCD07021501 CAS-Nummer: 886361-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...