Imidazo[1,2-a]pyridin-5-carbonsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2505-1G
COBBHC-2505-1GEA 0 EUR
COBBHC-2505-1G
Imidazo[1,2-a]pyridin-5-carbonsäure Hydrochlorid
Imidazo[1,2-a]pyridin-5-carbonsäure Hydrochlorid
Formel: C₈H₇ClN₂O₂
MDL: MFCD09954931
CAS-Nummer: 1198283-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR