3-(3-Bromphenyl)-5-(dichlormethyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-2518-1G
COBBHC-2518-1GEA 0 EUR
COBBHC-2518-1G
3-(3-Bromphenyl)-5-(dichlormethyl)-1,2,4-oxadiazol
3-(3-Bromophenyl)-5-(dichloromethyl)-1,2,4-oxadiazole
Formel: C₉H₅BrCl₂N₂O
MDL: MFCD12026023
CAS-Nummer: 1150164-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR