Ethyl-5-amino-4-cyan-1-(3-iodphenyl)-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-2530-1G
COBBHC-2530-1GEA 864 EUR
COBBHC-2530-1G
Ethyl-5-amino-4-cyan-1-(3-iodphenyl)-1H-pyrazol-3-carboxylat
Ethyl-5-amino-4-cyan-1-(3-iodphenyl)-1H-pyrazol-3-carboxylat
Formel: C₁₃H₁₁IN₄O₂
MDL: MFCD09835744
CAS-Nummer: 1150164-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR