2-(4-Brom-3-methyl-5-(trifluormethyl)-1H-pyrazol-1-yl)-6-chlorpyridin
Lieferant: COMBI-BLOCKS
HC-2542-5G
HC-2542-1G
COBBHC-2542-5GEA
303
EUR
COBBHC-2542-5G
COBBHC-2542-1G
2-(4-Brom-3-methyl-5-(trifluormethyl)-1H-pyrazol-1-yl)-6-chlorpyridin
2-(4-Brom-3-methyl-5-(trifluormethyl)-1H-pyrazol-1-yl)-6-chlorpyridin
Formel:
C₁₀H₆BrClF₃N₃ |
MDL:
MFCD12546466 CAS-Nummer: 1187386-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...