2,2-Difluor-2-(2-(trifluormethoxy)phenyl)essigsäure

Lieferant: COMBI-BLOCKS

HC-2556-1G
COBBHC-2556-1GEA 404 EUR
COBBHC-2556-1G
2,2-Difluor-2-(2-(trifluormethoxy)phenyl)essigsäure
2,2-Difluor-2-(2-(trifluormethoxy)phenyl)essigsäure
Formel: C₉H₅F₅O₃
Molecular Weight: 256,1262 g/mol
MDL: MFCD11855928
CAS-Nummer: 1133116-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR