2-(3-Brom-4-(trifluormethoxy)phenyl)-2,2-difluoressigsäure

Lieferant: COMBI-BLOCKS

HC-2558-1G
COBBHC-2558-1GEA 0 EUR
COBBHC-2558-1G
2-(3-Brom-4-(trifluormethoxy)phenyl)-2,2-difluoressigsäure
2-(3-Brom-4-(trifluormethoxy)phenyl)-2,2-difluoressigsäure
Formel: C₉H₄BrF₅O₃
MDL: MFCD11855929
CAS-Nummer: 1133116-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR