6-Brom-2-methoxy-4-methylchinolin

Lieferant: COMBI-BLOCKS

HC-2574-1G
COBBHC-2574-1GEA 595 EUR
COBBHC-2574-1G
6-Brom-2-methoxy-4-methylchinolin
6-Bromo-2-methoxy-4-methylquinoline
Formel: C₁₁H₁₀BrNO
MDL: MFCD12756430
CAS-Nummer: 1187386-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR