2-Brom-6-(cyclohexylamino)pyridin

Lieferant: COMBI-BLOCKS

HC-2723-5G HC-2723-1G
COBBHC-2723-5GEA 983 EUR
COBBHC-2723-5G COBBHC-2723-1G
2-Brom-6-(cyclohexylamino)pyridin
2-Brom-6-(cyclohexylamino)pyridin
Formel: C₁₁H₁₅BrN₂
MDL: MFCD09878953
CAS-Nummer: 959237-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR