2-Brom-6-(cyclohexylamino)pyridin
Lieferant: COMBI-BLOCKS
HC-2723-5G
HC-2723-1G
COBBHC-2723-5GEA
983
EUR
COBBHC-2723-5G
COBBHC-2723-1G
2-Brom-6-(cyclohexylamino)pyridin
2-Brom-6-(cyclohexylamino)pyridin
Formel:
C₁₁H₁₅BrN₂ |
MDL:
MFCD09878953 CAS-Nummer: 959237-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...