5-Chlor-2-methyl-4-thiocyanatoanilin

Lieferant: COMBI-BLOCKS

HC-2726-5G HC-2726-1G
COBBHC-2726-5GEA 227 EUR
COBBHC-2726-5G COBBHC-2726-1G
5-Chlor-2-methyl-4-thiocyanatoanilin
5-Chlor-2-methyl-4-thiocyanatoanilin
Formel: C₈H₇ClN₂S
MDL: MFCD12026046
CAS-Nummer: 23530-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR