Methyl-1H-indazol-7-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 1H-indazole-7-carboxylate

HC-2753-1G
COBBHC-2753-1GEA 50.9 EUR
COBBHC-2753-1G
Methyl-1H-indazol-7-carboxylat
Methyl-1H-indazol-7-carboxylat
Formel: C₉H₈N₂O₂
Molecular Weight: 176,1720 g/mol
MDL: MFCD07371608
CAS-Nummer: 755752-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR