4-Chlor-8-methyl-2-(trifluormethyl)chinolin
Lieferant: COMBI-BLOCKS
HC-2765-1G
COBBHC-2765-1GEA
303
EUR
COBBHC-2765-1G
4-Chlor-8-methyl-2-(trifluormethyl)chinolin
4-Chloro-8-methyl-2-(trifluoromethyl)quinoline
Formel:
C₁₁H₇ClF₃N |
MDL:
MFCD08448258 CAS-Nummer: 1701-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...