N-Methyl-3-bromanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2799-5G HC-2799-1G
COBBHC-2799-5GEA 121 EUR
COBBHC-2799-5G COBBHC-2799-1G
N-Methyl-3-bromanilin Hydrochlorid
N-Methyl-3-bromanilin Hydrochlorid
Formel: C₇H₉BrClN
MDL: MFCD12756434
CAS-Nummer: 152814-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR