2-(4-Methylcyclohexyliden)essigsäure

Lieferant: COMBI-BLOCKS

HC-2814-1G HC-2814-5G
COBBHC-2814-1GEA 98.3 EUR
COBBHC-2814-1G COBBHC-2814-5G
2-(4-Methylcyclohexyliden)essigsäure
2-(4-Methylcyclohexyliden)essigsäure
Formel: C₉H₁₄O₂
MDL: MFCD00029539
CAS-Nummer: 77842-31-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR