5'-Brom-2'-methylacetanilid

Lieferant: COMBI-BLOCKS

HC-2832-5G HC-2832-1G
COBBHC-2832-5GEA 0 EUR
COBBHC-2832-5G COBBHC-2832-1G
5'-Brom-2'-methylacetanilid
5'-Brom-2'-methylacetanilid
Formel: C₉H₁₀BrNO
MDL: MFCD11520466
CAS-Nummer: 116436-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR