4-Brom-2-nitro-5-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

HC-2840-1G HC-2840-5G
COBBHC-2840-1GEA 0 EUR
COBBHC-2840-1G COBBHC-2840-5G
4-Brom-2-nitro-5-(trifluormethyl)anilin
4-Brom-2-nitro-5-(trifluormethyl)anilin
Formel: C₇H₄BrF₃N₂O₂
MDL: MFCD09263997
CAS-Nummer: 683241-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR