Methyl-1H-indazol-6-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 1H-indazole-6-carboxylate, Indazole-6-carboxylic acid methyll ester, Methyl Indazole-6-carboxylate

HC-2847-1G HC-2847-5G
COBBHC-2847-1GEA 21.9 EUR
COBBHC-2847-1G COBBHC-2847-5G
Methyl-1H-indazol-6-carboxylat
Methyl-1H-indazol-6-carboxylat
Formel: C₉H₈N₂O₂
Molecular Weight: 176.17 g/mol
MDL: MFCD07371612
CAS-Nummer: 170487-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR