Methyl-1H-indazol-6-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 1H-indazole-6-carboxylate, Indazole-6-carboxylic acid methyll ester, Methyl Indazole-6-carboxylate
HC-2847-1G
HC-2847-5G
COBBHC-2847-1GEA
21.9
EUR
COBBHC-2847-1G
COBBHC-2847-5G
Methyl-1H-indazol-6-carboxylat
Methyl-1H-indazol-6-carboxylat
Formel:
C₉H₈N₂O₂ Molecular Weight: 176.17 g/mol |
MDL:
MFCD07371612 CAS-Nummer: 170487-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...