N-Methyl-2,3-dichloranilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2857-1G
COBBHC-2857-1GEA 303 EUR
COBBHC-2857-1G
N-Methyl-2,3-dichloranilin Hydrochlorid
N-Methyl-2,3-dichloranilin Hydrochlorid
Formel: C₇H₈Cl₃N
MDL: MFCD12912804
CAS-Nummer: 1187386-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR