3-Chlor-2-fluor-N-methylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: N-Methyl 3-chloro-2-fluoroaniline hydrochloride

HC-2858-1G
COBBHC-2858-1GEA 353 EUR
COBBHC-2858-1G
3-Chlor-2-fluor-N-methylanilin Hydrochlorid
3-Chlor-2-fluor-N-methylanilin Hydrochlorid
Formel: C₇H₈Cl₂FN
MDL: MFCD12913956
CAS-Nummer: 1187386-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR