5-Fluor-2-(methylamino)benzotrifluorid Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2880-1G HC-2880-5G
COBBHC-2880-1GEA 303 EUR
COBBHC-2880-1G COBBHC-2880-5G
5-Fluor-2-(methylamino)benzotrifluorid Hydrochlorid
5-Fluor-2-(methylamino)benzotrifluorid Hydrochlorid
Formel: C₈H₈ClF₄N
MDL: MFCD13195690
CAS-Nummer: 1215205-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR