2-Brom-5-iod-4-nitrobenzotrifluorid

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-4-iodo-5-nitro-2-(trifluoromethyl)benzene

HC-2983-1G
COBBHC-2983-1GEA 204 EUR
COBBHC-2983-1G
2-Brom-5-iod-4-nitrobenzotrifluorid
2-Brom-5-iod-4-nitrobenzotrifluorid
Formel: C₇H₂BrF₃INO₂
Molecular Weight: 395,9000 g/mol
MDL: MFCD12913962
CAS-Nummer: 1187385-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR