1-(3-Chlorphenyl)cyclopropancarbonsäure

Lieferant: COMBI-BLOCKS

HC-3004-1G
COBBHC-3004-1GEA 115 EUR
COBBHC-3004-1G
1-(3-Chlorphenyl)cyclopropancarbonsäure
1-(3-Chlorophenyl)cyclopropanecarboxylic acid
Formel: C₁₀H₉ClO₂
Molecular Weight: 196,6300 g/mol
MDL: MFCD07374442
CAS-Nummer: 124276-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR