3-Chlor-4-iodbenzotrifluorid

Lieferant: COMBI-BLOCKS

HC-3033-5G HC-3033-25G
COBBHC-3033-5GEA 37.4 EUR
COBBHC-3033-5G COBBHC-3033-25G
3-Chlor-4-iodbenzotrifluorid
3-Chlor-4-iodbenzotrifluorid
Formel: C₇H₃ClF₃I
Molecular Weight: 306.45 g/mol
MDL: MFCD00042562
CAS-Nummer: 141738-80-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR