7-Chlor-2-mercaptobenzothiazol

Lieferant: COMBI-BLOCKS

Synonyme: 7-chloro-3H-benzothiazole-2-thione

HC-3054-1G
COBBHC-3054-1GEA 363 EUR
COBBHC-3054-1G
7-Chlor-2-mercaptobenzothiazol
7-Chloro-2-mercaptobenzothiazole
Formel: C₇H₄ClNS₂
Molecular Weight: 201,7 g/mol
MDL: MFCD06408879
CAS-Nummer: 1849-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR