6-Chlorchinoxalin

Lieferant: COMBI-BLOCKS

HC-3083-5G HC-3083-25G HC-3083-1G
COBBHC-3083-5GEA 0 EUR
COBBHC-3083-5G COBBHC-3083-25G COBBHC-3083-1G
6-Chlorchinoxalin
6-Chloroquinoxaline
Formel: C₈H₅ClN₂
MDL: MFCD00047594
CAS-Nummer: 5448-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR