6-Chlor-7-methylchinoxalin

Lieferant: COMBI-BLOCKS

HC-3257-5G HC-3257-1G
COBBHC-3257-5GEA 451 EUR
COBBHC-3257-5G COBBHC-3257-1G
6-Chlor-7-methylchinoxalin
6-Chloro-7-methylquinoxaline
Formel: C₉H₇ClN₂
MDL: MFCD15143565
CAS-Nummer: 646504-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR