2-(2-Chlorphenyl)oxazol

Lieferant: COMBI-BLOCKS

HC-3301-5G HC-3301-1G
COBBHC-3301-5GEA 0 EUR
COBBHC-3301-5G COBBHC-3301-1G
2-(2-Chlorphenyl)oxazol
2-(2-Chlorophenyl)oxazole
Formel: C₉H₆ClNO
MDL: MFCD03407330
CAS-Nummer: 62881-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR