1-Brom-3-cyclopentenylbenzol

Lieferant: COMBI-BLOCKS

HC-3322-5G HC-3322-1G
COBBHC-3322-5GEA 451 EUR
COBBHC-3322-5G COBBHC-3322-1G
1-Brom-3-cyclopentenylbenzol
1-Brom-3-cyclopentenylbenzol
Formel: C₁₁H₁₁Br
MDL: MFCD16036124
CAS-Nummer: 19920-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR