2-(4-Nitrophenyl)oxazol

Lieferant: COMBI-BLOCKS

HC-3339-5G HC-3339-1G
COBBHC-3339-5GEA 593 EUR
COBBHC-3339-5G COBBHC-3339-1G
2-(4-Nitrophenyl)oxazol
2-(4-Nitrophenyl)oxazole
Formel: C₉H₆N₂O₃
MDL: MFCD08276758
CAS-Nummer: 62882-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR