4-(1,3-Oxazol-2-yl)anilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-3349-1G
COBBHC-3349-1GEA 376 EUR
COBBHC-3349-1G
4-(1,3-Oxazol-2-yl)anilin Hydrochlorid
4-(1,3-Oxazol-2-yl)anilin Hydrochlorid
Formel: C₉H₉ClN₂O
MDL: MFCD19237193
CAS-Nummer: 1351659-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR