2-Iod-4-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Iodo-1-methoxy-4-nitrobenzene

HC-3364-5G HC-3364-25G
COBBHC-3364-5GEA 0 EUR
COBBHC-3364-5G COBBHC-3364-25G
2-Iod-4-nitroanisol
2-Iod-4-nitroanisol
Formel: C₇H₆INO₃
MDL: MFCD00024328
CAS-Nummer: 5399-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR