2-Iod-4-(trifluormethyl)anilin
Lieferant: COMBI-BLOCKS
Synonyme:
2-Iod-α,α,α-trifluor-p-toluidin, 4-Amino-3-iod-α,α,α-trifluortoluol
HC-3375-25G
HC-3375-5G
COBBHC-3375-25GEA
101
EUR
COBBHC-3375-25G
COBBHC-3375-5G
2-Iod-4-(trifluormethyl)anilin
2-Iod-4-(trifluormethyl)anilin
Formel:
C₇H₅F₃IN Molecular Weight: 287.02 g/mol |
MDL:
MFCD02683097 CAS-Nummer: 163444-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...