2-Iod-4-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Iod-α,α,α-trifluor-p-toluidin, 4-Amino-3-iod-α,α,α-trifluortoluol

HC-3375-25G HC-3375-5G
COBBHC-3375-25GEA 101 EUR
COBBHC-3375-25G COBBHC-3375-5G
2-Iod-4-(trifluormethyl)anilin
2-Iod-4-(trifluormethyl)anilin
Formel: C₇H₅F₃IN
Molecular Weight: 287.02 g/mol
MDL: MFCD02683097
CAS-Nummer: 163444-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR