4-Chlor-2-iodanilin

Lieferant: COMBI-BLOCKS

HC-3380-25G
COBBHC-3380-25GEA 76.3 EUR
COBBHC-3380-25G
4-Chlor-2-iodanilin
4-Chlor-2-iodanilin
Formel: C₆H₅ClIN
Molecular Weight: 253,47 g/mol
MDL: MFCD01863737
CAS-Nummer: 63069-48-7
EINECS: 263-839-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR