5-Brom-2-iodanilin

Lieferant: COMBI-BLOCKS

HC-3389-1G
COBBHC-3389-1GEA 52.5 EUR
COBBHC-3389-1G
5-Brom-2-iodanilin
5-Brom-2-iodanilin
Formel: C₆H₅BrIN
Molecular Weight: 297,9200 g/mol
MDL: MFCD09753734
CAS-Nummer: 64085-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR