1-Brom-4-chlor-2-iodbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-5-chloroiodobenzene, 1-Bromo-4-Chloro-2-Iodobenzene

HC-3434-5G
COBBHC-3434-5GEA 154 EUR
COBBHC-3434-5G
1-Brom-4-chlor-2-iodbenzol
1-Bromo-4-chloro-2-iodobenzene
Formel: C₆H₃BrCl
Molecular Weight: 317,3495 g/mol
MDL: MFCD00672943
CAS-Nummer: 148836-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR