1-(2-Bromphenyl)pyrrolidin

Lieferant: COMBI-BLOCKS

HC-3496-1G
COBBHC-3496-1GEA 312 EUR
COBBHC-3496-1G
1-(2-Bromphenyl)pyrrolidin
1-(2-Bromphenyl)pyrrolidin
Formel: C₁₀H₁₂BrN
Molecular Weight: 226,1130 g/mol
MDL: MFCD20486327
CAS-Nummer: 87698-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR